Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Thymidylate Kinase/TMPK, Clone: 1A5.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC609
Description
Specifically detects Thymidylate Kinase/TMPK clone: 1A5.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Thymidylate kinase (EC 2.7.4.9; UniProt P23919; also known as dTMP kinase) is encoded by the DTYMK (also known as CDC8, PP3731, TMPK, TYMK) gene (Gene ID 1841) in human. Thymidylate kinase (TMPK) is an ubiquitously expressed enzyme of the dTTP synthesis pathway involved in DNA synthesis. TMPK catalyzes the phosphorylation of thymidine 5'-monophosphate (dTMP) in the presence of ATP and Mg2+ to form thymidine 5'-diphosphate (dTDP), which is then further phosphorylated by nucleoside-diphosphate kinase (NDK) to thymidine 5'-triphosphate (dTTP). Aside from its normal physiological role, TMPK activity is essential for the activation of nucleoside analogs-based prodrugs, such as the clinically used anti-HIV prodrugs AZT and d4T, which need to be converted to the triphosphate forms for their pharmacological activities. Inhibition of TMPK activity is reported to augment the potency of anticancer agent doxorubicin against human colon cancer cells regardless of their p53 status. In addition, lung cancers with LKB1-mutations are reported to show deficits in nucleotide metabolism and exhibit hypersensitivity to DTYMK inhibition.
Specifications
Thymidylate Kinase/TMPK | |
Monoclonal | |
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
DTYMK; CDC8; TMPK; TYMK | |
GST-tagged recombinant protein corresponding to human Thymidylate Kinase/TMPK. | |
100 μg | |
Apoptosis, Cancer | |
NP_001158503 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
1A5.1 | |
P23919 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction