Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid-stimulating hormone (TSH), Mouse, Clone: 2/3E1 2F12, Abnova™

Catalog No. 89135507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-507 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-507 Supplier Abnova Corporation Supplier No. MAB3699
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Thyroid-stimulating hormone (TSH).

Specifications

Antigen Thyroid-stimulating hormone (TSH)
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 2/3E1 2F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Thyroid-stimulating hormone (TSH).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Recombinant protein corresponding to human thyroid-stimulating hormone.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes natural and recombinant human Thyroid Stimulating Hormone.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.