Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid-stimulating hormone (TSH), Mouse, Clone: 204-12252, Abnova™

Catalog No. 89135759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-759 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-759 Supplier Abnova Corporation Supplier No. MAB4745
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Thyroid-stimulating hormone (TSH).

Specifications

Antigen Thyroid-stimulating hormone (TSH)
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 204-12252
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Thyroid-stimulating hormone (TSH).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Purified human TSH and TSH beta chain.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cross-reactivity: TSH alpha 1.0% TSH beta 0.6% FSH 0.6% LH 0.3%
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.