Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroxine, Mouse, Clone: XM212, Abnova™

Catalog No. 89135288 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-288 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-288 Supplier Abnova Corporation Supplier No. MAB2886
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Thyroxine.

Specifications

Antigen Thyroxine
Applications ELISA
Classification Monoclonal
Clone XM212
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Thyroxine.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM Tris, 150mM NaCl, pH 7.5 (0.05% sodium azide)
Host Species Mouse
Immunogen Thyroxine conjugated with BSA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.