Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroxine (T4), Mouse, Clone: ME.125, Abnova™

Catalog No. 89135762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-762 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-762 Supplier Abnova Corporation Supplier No. MAB4752

Mouse monoclonal antibody raised against Thyroxine (T4).

Specifications

Antigen Thyroxine (T4)
Applications ELISA
Classification Monoclonal
Clone ME.125
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Thyroxine (T4).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 150mM PBS, pH 7.4
Host Species Mouse
Immunogen conjugated via carboxy (-COOH) site of molecule., Thyroxine (T4)-BSA
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes free T4 and total thyroxine. Thyroxine (T4) a thyroid hormone exerts a potent influence on the basal metabolic rate. Synthesized from the tyrosine residues of thyroglobin and is found in the serum of normal individuals. The L-form is twice as active physiologically as a racemic product, the D-form has very little activity. Cross Reactivity: <1% for T3, T2, and rT3 by direct EIA.
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.