Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tlr12, Mouse, Clone: 15F1215, Abnova™

Catalog No. 89107872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-872 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-872 Supplier Abnova Corporation Supplier No. MAB0104
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Tlr12.

Sequence: LGLRLCLPERDFEPGK

Specifications

Antigen Tlr12
Applications Western Blot
Classification Monoclonal
Clone 15F1215
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Tlr12.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Tlr12
Gene Alias Gm1365
Gene Symbols Tlr12
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 791-806 of mouse Tlr12.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 384059
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.