Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 isoform A, Mouse, Clone: 26C593.2, Abnova™

Catalog No. 89107858 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-858 Supplier Abnova Corporation Supplier No. MAB0087
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TLR9.

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is preferentially expressed in immune cell rich tissues, such as spleen, lymph node, bone marrow and peripheral blood leukocytes. Studies in mice and human indicate that this receptor mediates cellular response to unmethylated CpG dinucleotides in bacterial DNA to mount an innate immune response. [provided by RefSeq

Specifications

Antigen TLR9 isoform A
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 26C593.2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TLR9.
Dilution ELISA (1:100-1:2000) Flow Cytometry Immunocytochemistry (1:10-1:500) Immunofluorescence (1:10-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation (1:10-1:500) Western Blot (2-5 ug/mL)
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene TLR9
Gene Alias CD289
Gene Symbols TLR9
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 268-300 of human TLR9 isoform A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54106
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.