Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMBIM6, Mouse, Clone: 20F248, Abnova™

Catalog No. 89107874 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-874 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-874 Supplier Abnova Corporation Supplier No. MAB0106
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TMBIM6.

Specifications

Antigen TMBIM6
Applications Western Blot
Classification Monoclonal
Clone 20F248
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TMBIM6.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene TMBIM6
Gene Alias BAXI1/BI-1/TEGT
Gene Symbols TMBIM6
Host Species Mouse
Immunogen A mixture of synthetic peptides corresponding to N-terminus, internal and C-terminuss of human TMBIM6.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7009
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.