Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM81, Mouse, Clone: 3D5, Abnova™

Catalog No. 89107789 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-789 Supplier Abnova Corporation Supplier No. H00388730M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TMEM81.

Sequence: MKVLATSFVLGSLGLAFYLPLVVTTPKTLAIPEKLQEAVGKVIINATTCTVTCGLGYKEETVCEVGPDGVRRKCQTRRLECLTNWICGMLHFTILIGKEFELSCLSSDILEFGQEAFRFTWRLARGVISTDDEVFKPFQANSHFVKFKYAQEYDSGTYRCDVQLVKNLRLVKRLYFGLRVLPPNLVNLNFHQSLTEDQKLIDEGLEVNLDSYSKPHHPKWKKKVASALGIGIAIGVVGGVLVRIVLCALRGGLQQ

Specifications

Antigen TMEM81
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant TMEM81.
Formulation PBS with no preservative; pH 7.4
Gene TMEM81
Gene Accession No. NM_203376.1
Gene Alias HC3107/KVLA2788/MGC75217/UNQ2788
Gene Symbols TMEM81
Host Species Mouse
Immunogen TMEM81 (NP_976310.1,1 a.a. ∽ 255 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388730
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.