Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF, Mouse, Clone: CH8820, Abnova™

Catalog No. 89112825 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-825 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-825 Supplier Abnova Corporation Supplier No. MAB6323
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TNF.

This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophages. It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. Knockout studies in mice also suggested the neuroprotective function of this cytokine. [provided by RefSeq

Specifications

Antigen TNF
Applications Immunohistochemistry, Inhibition Assays, ELISA
Classification Monoclonal
Clone CH8820
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant TNF.
Dilution Sandwich ELISA (Capture Coating Dose: 1.0-2.0 ug/well) Inhibition Assay (2-5 ng of rTNF-alpha per ug) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.0
Gene TNF
Gene Alias DIF/TNF-alpha/TNFA/TNFSF2
Gene Symbols TNF
Host Species Mouse
Immunogen Recombinant protein corresponding to human TNF.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity This antibody recognizes both, recombinant and native human TNF.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.