Learn More
TNF, Mouse, Clone: TA70, Abnova™
Mouse monoclonal antibody raised against recombinant TNF.
Supplier: Abnova Corporation MAB3052
Description
This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophages. It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. Knockout studies in mice also suggested the neuroprotective function of this cytokine. [provided by RefSeq
Specifications
TNF | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
TNF | |
TNF | |
Recombinant protein corresponding to human TNF. | |
RUO | |
7124 | |
Human | |
Liquid |
ELISA | |
TA70 | |
Mouse monoclonal antibody raised against recombinant TNF. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
DIF/TNF-alpha/TNFA/TNFSF2 | |
Mouse | |
200 μg | |
Primary | |
This antibody is specific to TNF. | |
Store at 4°C. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.