Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-TNFα Receptor, Clone: 13F9.1,

Catalog No. MABC615MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC615MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC615MI Supplier MilliporeSigma™ Supplier No. MABC615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TNFα Receptor clone: 13F9.1 in Human, Mouse samples, and it is validated for Immunohistochemistry, Western Blotting

Tumor necrosis factor receptor superfamily member 1A (UniProt P19438; also known as CD120a, p55, p60, TNF-R1, TNF-RI, TNFR-I, Tumor necrosis factor-alpha receptor, Tumor necrosis factor receptor type 1) is encoded by the TNFRSF1A (also known as FPF, MS5, TNFAR, TNFAR55, TNFAR60, TNFR1) gene (Gene ID 7132) in human. It functions as a receptor for TNFSF2/TNF-alpha and homotrimeric TNFSF1/lymphotoxin-alpha. Upon activation, TNFalpha receptor mediates apoptotic, anti-viral, and other cellular signaling events. Binding of TNF-alpha to the extracellular domain of TNF receptor leads to homotrimerization. Proteolytic cleavage results in the relese of Tumor necrosis factor-binding protein 1 (TBP1). Defects in TNF receptor cause tumor necrosis factor receptor-associated periodic syndrome (TRAPS), a hereditary periodic fever syndrome characterized by recurrent fever, abdominal pain, localized tender skin lesions and myalgia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFα Receptor
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 13F9.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P19438
Gene Symbols TNFRSF1A; TNFAR; TNFR1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the extracellular domain of human TNFα Receptor.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001056
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.