Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP3, Mouse, Clone: 8E8.38, Abnova™

Catalog No. 89111557 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-557 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-557 Supplier Abnova Corporation Supplier No. MAB3716

Mouse monoclonal antibody raised against partial TNFAIP3.

This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses. [provided by RefSeq

Specifications

Antigen TNFAIP3
Applications Western Blot
Classification Monoclonal
Clone 8E8.38
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial TNFAIP3.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene TNFAIP3
Gene Alias A20/MGC104522/MGC138687/MGC138688/OTUD7C/TNFA1P2
Gene Symbols TNFAIP3
Host Species Mouse
Immunogen Zinc finger domain (amino acids 375-537) of human TNFAIP3.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7128
Test Specificity Recognizes human A20.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.