Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF10A, Mouse, FITC, Clone: DR-4-02, Abnova™

Catalog No. 89112099 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-099 Supplier Abnova Corporation Supplier No. MAB5076
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TNFRSF10A.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL), and thus transduces cell death signal and induces cell apoptosis. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by this protein. [provided by RefSeq

Specifications

Antigen TNFRSF10A
Applications Flow Cytometry
Classification Monoclonal
Clone DR-4-02
Concentration 0.1 mg/mL
Conjugate FITC
Description Mouse monoclonal antibody raised against partial recombinant TNFRSF10A.
Dilution Flow Cytometry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TNFRSF10A
Gene Alias APO2/CD261/DR4/MGC9365/TRAILR-1/TRAILR1
Gene Symbols TNFRSF10A
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to extracellular region of TNFRSF10A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8797
Test Specificity This antibody recognizes TRAIL-R1 (DR4), a human death receptor 4 (468 amino acids) expressed in most human tissues (spleen, peripheral blood leucocytes, thymus) and in a variety of tumour-derived.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.