Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF10C, Mouse, PE, Clone: TRAIL-R3-02, Abnova™

Catalog No. 89112142 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-142 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-142 Supplier Abnova Corporation Supplier No. MAB5138
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TNFRSF10C.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain and a transmembrane domain, but no cytoplasmic death domain. This receptor is not capable of inducing apoptosis, and is thought to function as an antagonistic receptor that protects cells from TRAIL-induced apoptosis. This gene was found to be a p53-regulated DNA damage-inducible gene. The expression of this gene was detected in many normal tissues but not in most cancer cell lines, which may explain the specific sensitivity of cancer cells to the apoptosis-inducing activity of TRAIL. [provided by RefSeq

Specifications

Antigen TNFRSF10C
Applications Flow Cytometry
Classification Monoclonal
Clone TRAIL-R3-02
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against recombinant TNFRSF10C.
Dilution Flow Cytometry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TNFRSF10C
Gene Alias CD263/DCR1/LIT/MGC149501/MGC149502/TRAILR3/TRID
Gene Symbols TNFRSF10C
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to amino acids 1-280 of TNFRSF10C.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8794
Test Specificity This antibody reacts with TRAIL-R3, a 35 KDa GPI-anchored extracellular membrane protein expressed mainly on neutrophils.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.