Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF10D, Mouse, FITC, Clone: TRAIL-R4-01, Abnova™

Catalog No. 89112143 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-143 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-143 Supplier Abnova Corporation Supplier No. MAB5139
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TNFRSF10D.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain, a transmembrane domain, and a truncated cytoplamic death domain. This receptor does not induce apoptosis, and has been shown to play an inhibitory role in TRAIL-induced cell apoptosis. [provided by RefSeq

Specifications

Antigen TNFRSF10D
Applications Flow Cytometry
Classification Monoclonal
Clone TRAIL-R4-01
Concentration 0.1 mg/mL
Conjugate FITC
Description Mouse monoclonal antibody raised against partial recombinant TNFRSF10D.
Dilution Flow Cytometry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TNFRSF10D
Gene Alias CD264/DCR2/TRAILR4/TRUNDD
Gene Symbols TNFRSF10D
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to amino acids 1-210 of TNFRSF10D.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8793
Test Specificity This antibody reacts with TRAIL-R4, a 42 KDa transmembrane protein expressed on various blood cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.