Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF21, Mouse, PE, Clone: DR-6-04-EC, Abnova™

Catalog No. 89111844 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-844 Supplier Abnova Corporation Supplier No. MAB4474

Mouse monoclonal antibody raised against partial recombinant TNFRSF21.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors. Knockout studies in mice suggested that this gene plays a role in T-helper cell activation, and may be involved in inflammation and immune regulation. [provided by RefSeq

Specifications

Antigen TNFRSF21
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone DR-6-04-EC
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against partial recombinant TNFRSF21.
Dilution Flow Cytometry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TNFRSF21
Gene Alias BM-018/DR6/MGC31965
Gene Symbols TNFRSF21
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to amino acids 42-335 extracellular region of human TNFRSF21.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27242
Test Specificity This antibody recognizes human DR6 (Death receptor-6), a type I transmembrane protein containing cytoplasmic death domain, widely expressed in most human tissues and cell lines.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.