Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF8, Mouse, PE, Clone: MEM-268, Abnova™

Catalog No. 89112114 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-114 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-114 Supplier Abnova Corporation Supplier No. MAB5100
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against TNFRSF8.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is expressed by activated, but not by resting, T and B cells. TRAF2 and TRAF5 can interact with this receptor, and mediate the signal transduction that leads to the activation of NF-kappaB. This receptor is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Specifications

Antigen TNFRSF8
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-268
Conjugate PE
Description Mouse monoclonal antibody raised against TNFRSF8.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TNFRSF8
Gene Alias CD30/D1S166E/KI-1
Gene Symbols TNFRSF8
Host Species Mouse
Immunogen Expression vector containing CD30 cDNA (booster suspension of THP-1 cell line).
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Test Specificity This antibody recognizes extracellular part of CD30 (Ki-1 antigen), a 105 KDa single chain glycoprotein expressed on Hodgkin's and Reed-Sternberg cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.