Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF10, Mouse, Clone: SB91c1, Abnova™

Catalog No. 89113386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-386 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-386 Supplier Abnova Corporation Supplier No. MAB8219
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant TNFSF10.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. This protein binds to several members of TNF receptor superfamily including TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and possibly also to TNFRSF11B/OPG. The activity of this protein may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and TNFRSF11B/OPG that cannot induce apoptosis. The binding of this protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3. [provided by RefSeq

Specifications

Antigen TNFSF10
Applications ELISA
Classification Monoclonal
Clone SB91c
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant TNFSF10.
Dilution ELISA (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.0
Gene TNFSF10
Gene Alias APO2L/Apo-2L/CD253/TL2/TRAIL
Gene Symbols TNFSF10
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CASP8.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8743
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.