Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF13B, Mouse, Clone: ANC2H3, Abnova™

Catalog No. 89113107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-107 Supplier Abnova Corporation Supplier No. MAB6983
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TNFSF13B.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells. [provided by RefSeq

Specifications

Antigen TNFSF13B
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone ANC2H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant TNFSF13B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate)
Gene TNFSF13B
Gene Alias BAFF/BLYS/CD257/DTL/TALL-1/TALL1/THANK/TNFSF20/ZTNF4
Gene Symbols TNFSF13B
Host Species Mouse
Immunogen Recombinant protein corresponding to human TNFSF13B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Test Specificity Antibody ANC2H3 blocks binding of recombinant CD257 to Raji cell surface.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.