Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNN, Mouse, Clone: T20, Abnova™

Catalog No. 89110477 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-477 Supplier Abnova Corporation Supplier No. MAB1529
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TNN.

Specifications

Antigen TNN
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone BC-24
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TNN.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) Immunohistochemistry (Frozen sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene TNN
Gene Alias TN-W
Gene Symbols TNN
Host Species Mouse
Immunogen Native purified human TNN.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63923
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.