Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNNI3, Mouse, Clone: 3A10A12, 5C3A7, Abnova™

Catalog No. 89113423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-423 Supplier Abnova Corporation Supplier No. MAB8299
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TNNI3.

Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. This gene encodes the TnI-cardiac protein and is exclusively expressed in cardiac muscle tissues. Mutations in this gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM). [provided by RefSeq

Specifications

Antigen TNNI3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 3A10A12, 5C3A7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TNNI3.
Dilution Western Blot (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:1000) Immunohistochemistry (Frozen sections) (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene TNNI3
Gene Alias CMD2A/CMH7/MGC116817/RCM1/TNNC1/cTnI
Gene Symbols TNNI3
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human TNNI3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7137
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.