Learn More
TNNT2, Mouse, Clone: 7E7, Abnova™
Mouse monoclonal antibody raised against TNNT2.
Supplier: Abnova Corporation MAB3100
Description
The protein encoded by this gene is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Mutations in this gene have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for this gene undergo alternative splicing that results in many tissue-specific isoforms, however, the full-length nature of some of these variants has not yet been determined. [provided by RefSeq
Specifications
TNNT2 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
TNNT2 | |
TNNT2 | |
Human TNNT2 complex. | |
RUO | |
7139 | |
Store at 4°C. | |
IgG |
ELISA | |
7E7 | |
Mouse monoclonal antibody raised against TNNT2. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
CMH2/CMPD2/MGC3889/RCM3/TnTC/cTnT | |
Mouse | |
200 μg | |
Primary | |
Bovine, Canine, Human, Monkey, Mouse, Pig, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.