Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNNT2, Mouse, Clone: 7G7, Abnova™

Catalog No. 89111242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-242 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-242 Supplier Abnova Corporation Supplier No. MAB3094
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against TNNT2.

The protein encoded by this gene is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Mutations in this gene have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for this gene undergo alternative splicing that results in many tissue-specific isoforms, however, the full-length nature of some of these variants has not yet been determined. [provided by RefSeq

Specifications

Antigen TNNT2
Applications ELISA
Classification Monoclonal
Clone 7G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against TNNT2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TNNT2
Gene Alias CMH2/CMPD2/MGC3889/RCM3/TnTC/cTnT
Gene Symbols TNNT2
Host Species Mouse
Immunogen Human TNNT2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7139
Test Specificity This antibody has no cross-reaction with TNNI1.
Target Species Bovine, Canine, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.