Learn More
TNPO1, Mouse, Clone: D45, Abnova™
Mouse monoclonal antibody raised against recombinant TNPO1.
Supplier: Abnova Corporation MAB2496
Description
This gene encodes the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins. Alternate splicing of this gene results in two transcript variants encoding different proteins. [provided by RefSeq
Specifications
TNPO1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
TNPO1 | |
TNPO1 | |
Recombinant His fusion protein corresponding to human TNPO1. | |
RUO | |
3842 | |
Human, Monkey, Rabbit | |
Liquid |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
D45 | |
Mouse monoclonal antibody raised against recombinant TNPO1. | |
In PBS (0.09% sodium azide) | |
IPO2/KPNB2/MIP/MIP1/TRN | |
Mouse | |
100 μL | |
Primary | |
Detects a band of approximately 90 KDa. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.