Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNPO1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-818 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-818 Supplier Abnova Corporation Supplier No. H00003842A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TNPO1.

This gene encodes the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins. Alternate splicing of this gene results in two transcript variants encoding different proteins. [provided by RefSeq

Sequence: PDTTIQRTVQQKLEQLNQYPDFNNYLIFVLTKLKSEDEPTRSLSGLILKNNVKAHFQNFPNGVTDFIKSECLNNIGDSSPLIRATVGILITTIASKGELQNWPDLLPKLCSLLDSED

Specifications

Antigen TNPO1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TNPO1.
Formulation 50% glycerol
Gene TNPO1
Gene Accession No. NM_002270
Gene Alias IPO2/KPNB2/MIP/MIP1/TRN
Gene Symbols TNPO1
Host Species Mouse
Immunogen TNPO1 (NP_002261, 25 a.a. ∽ 141 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3842
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.