Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP73, Mouse, Clone: 24, Abnova™

Catalog No. 89113684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-684 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-684 Supplier Abnova Corporation Supplier No. MAB9549

Mouse monoclonal antibody raised against partial recombinant TP73.

This gene encodes tumor protein p73, which is a member of the p53 family of transcription factors involved in cellular responses to stress and development. The family members include p53, p63, and p73 and have high sequence similarity to one another, which allows p63 and p73 to transactivate p53-responsive genes causing cell cycle arrest and apoptosis. The family members can interact with each other in many ways involving direct or indirect protein interactions, resulting in regulation of the same target gene promoters or regulation of each other's promoters. The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. The p73 gene expresses at least 35 mRNA variants due to the use of alternate promoters, alternate translation initiation sites, and multiple splice variations. Theoretically this can account for 29 different p73 isoforms; however, the biological validity and the full-length nature of most variants have not been determined. [provided by RefSeq]

Specifications

Antigen TP73
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 24
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TP73.
Dilution Immunohistochemistry (Frozen sections) (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(1:50) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene TP73
Gene Alias P73
Gene Symbols TP73
Host Species Mouse
Immunogen Recombinant protein corresponding to 110 amino acids at C-terminus of human TP73.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.