Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52L1, Mouse, Clone: d1C5, Abnova™

Catalog No. 89108696 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-696 Supplier Abnova Corporation Supplier No. MAB1051
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TPD52L1.

This gene encodes a member of the tumor protein D52 (TPD52) family. The encoded protein contains a coiled-coil domain and may form homo- or hetero-dimer with TPD52 family members. The protein is reported to be involved in cell proliferation and calcium signaling. It also interacts with the mitogen-activated protein kinase kinase kinase 5 (MAP3K5/ASK1) and positively regulates MAP3K5-induced apoptosis. Multiple alternatively spliced transcript variants have been observed, but the full-length nature of some variants has not yet been determined. [provided by RefSeq

Specifications

Antigen TPD52L1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone d1C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TPD52L1.
Dilution ELISA Flow Cytometry Immunohistochemistry Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TPD52L1
Gene Accession No. NM_001003397
Gene Alias D53/MGC8556/TPD52L2/hD53
Gene Symbols TPD52L1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-131 of human TPD52L1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7164
Target Species Human
Content And Storage Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.