Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transmissible gastroenteritis virus, Mouse, Clone: TGEV11-314.6, Abnova™

Catalog No. 89136457 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-136-457 Supplier Abnova Corporation Supplier No. MAB8325
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against Transmissible gastroenteritis virus.

Specifications

Antigen Transmissible gastroenteritis virus
Applications ELISA
Classification Monoclonal
Clone TGEV11-314.6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Transmissible gastroenteritis virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Native purified from Transmissible gastroenteritis virus-infected ST cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies specifically recognize TGEV and do not cross-react with the following coronaviruses: FIPV, FeCV, CCV.
Target Species Virus
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.