Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAT1, Mouse, PE, Clone: TRIM-04, Abnova™

Catalog No. 89111872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-872 Supplier Abnova Corporation Supplier No. MAB4526
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TRAT1.

Specifications

Antigen TRAT1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TRIM-04
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against partial recombinant TRAT1.
Dilution Flow Cytometry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene TRAT1
Gene Alias HSPC062/TCRIM/TRIM
Gene Symbols TRAT1
Host Species Mouse
Immunogen Recombinant protein corresponding to partial human TRAT1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50852
Test Specificity This antibody reacts with T cell receptor-interacting molecule (TRIM), a 30 KDa adaptor protein expressed by T cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.