Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM2, Mouse, Clone: 2B5, Abnova™

Catalog No. 89110797 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-797 Supplier Abnova Corporation Supplier No. MAB2056

Mouse monoclonal antibody raised against partial recombinant TREM2.

The protein encoded by this gene is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL). [provided by RefSeq

Specifications

Antigen TREM2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TREM2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TREM2
Gene Alias TREM-2/Trem2a/Trem2b/Trem2c
Gene Symbols TREM2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 19-161 of human TREM2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54209
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.