Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM16, Mouse, Clone: 5G11, Abnova™

Catalog No. 89106372 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-372 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-372 Supplier Abnova Corporation Supplier No. H00010626M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRIM16.

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined. [provided by RefSeq

Sequence: LDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMA

Specifications

Antigen TRIM16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRIM16.
Formulation ascites with no preservative
Gene TRIM16
Gene Accession No. NM_006470
Gene Alias EBBP
Gene Symbols TRIM16
Host Species Mouse
Immunogen TRIM16 (NP_006461,165 a.a. ∽ 273 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10626
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.