Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM39, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-141 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-141 Supplier Abnova Corporation Supplier No. H00056658A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TRIM39.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: TPLPETGYWRVRLWNGDKYAATTTPFTPLHIKVKPKRVGIFLDYEAGTLSFYNVTDRSHIYTFTDTFTEKLWPLFYPGIRAGRKNAAPLTIRPPTDW

Specifications

Antigen TRIM39
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TRIM39.
Formulation 50% glycerol
Gene TRIM39
Gene Accession No. NM_021253
Gene Alias MGC32984/RNF23/TFP/TRIM39B
Gene Symbols TRIM39
Host Species Mouse
Immunogen TRIM39 (NP_067076, 421 a.a. ∽ 517 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56658
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.