Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM5, Mouse, Clone: 3B11H2, Abnova™

Catalog No. 89112294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-294 Supplier Abnova Corporation Supplier No. MAB5551
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TRIM5.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Five alternatively spliced transcript variants for this gene have been described. However, the full length nature of two of the variants has not been determined. [provided by RefSeq

Specifications

Antigen TRIM5
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 3B11H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TRIM5.
Dilution Western Blot (1:1000-1:5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:400) Immunohistochemistry (Frozen sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene TRIM5
Gene Alias RNF88/TRIM5alpha
Gene Symbols TRIM5
Host Species Mouse
Immunogen Recombinant protein corresponding to human TRIM5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.