Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM63, Mouse, Clone: 6G6, Abnova™

Catalog No. 89107459 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-459 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-459 Supplier Abnova Corporation Supplier No. H00084676M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRIM63.

This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle. [provided by RefSeq]

Sequence: DKSTKLVETAIQSLDEPGGATFLLTAKQLIKSIVEASKGCQLGKTEQGFENMDFFTLDLEHIADALRAIDFGTDEEEEEFIEEEDQEEEESTEGKEEGH

Specifications

Antigen TRIM63
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRIM63.
Formulation ascites with no preservative
Gene TRIM63
Gene Accession No. NM_032588
Gene Alias FLJ32380/IRF/MURF1/MURF2/RNF28/SMRZ
Gene Symbols TRIM63
Host Species Mouse
Immunogen TRIM63 (NP_115977,254 a.a. ∽ 352 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84676
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.