Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trp53, Mouse, Clone: Pab122, Abnova™

Catalog No. 89110737 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-737 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-737 Supplier Abnova Corporation Supplier No. MAB1970
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Trp53.

Specifications

Antigen Trp53
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Pab122
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Trp53.
Dilution Western blot (5 to 10 ug/mL) Immunoprecipitation (2 ug/mg) Immunohistochemistry on frozen formalin to fixed/paraffin to embedded tissues (1 to 2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Trp53
Gene Alias bbl/bfy/bhy/p53
Gene Symbols Trp53
Host Species Mouse
Immunogen Native purified human Trp53.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22059
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.