Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC6, Mouse, Clone: 3F2.H10.F2, Abnova™

Catalog No. 89113370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-370 Supplier Abnova Corporation Supplier No. MAB8167
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against synthetic peptide of TRPC6 .

The protein encoded by this gene forms a receptor-activated calcium channel in the cell membrane. The channel is activated by diacylglycerol and is thought to be under the control of a phosphatidylinositol second messenger system. Activation of this channel occurs independently of protein kinase C and is not triggered by low levels of intracellular calcium. Defects in this gene are a cause of focal segmental glomerulosclerosis 2 (FSGS2). [provided by RefSeq

Specifications

Antigen TRPC6
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F2.H10.F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TRPC6 .
Dilution ELISA (1:10000-1:50000) Western Blot (1:500-1:2000) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene TRPC6
Gene Alias FLJ11098/FLJ14863/FSGS2/TRP6
Gene Symbols TRPC6
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human TRPC6 .
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7225
Target Species Chimpanzee, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.