Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-TRPM1, Clone: 545H5,

Catalog No. MABN1841 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1841 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1841 Supplier MilliporeSigma™ Supplier No. MABN1841
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TRPM1 clone: 545H5 in Mouse samples, and it is validated for Western Blotting

Transient receptor potential cation channel subfamily M member 1 (UniProt Q2TV84; also known as Long transient receptor potential channel 1, LTrpC1, Melastatin-1) is encoded by the Trpm1 (also known as CSNB1C, Ltrpc1, Mlsn1, MLSN1) gene (Gene ID 17364) in murine species. TRPM1 belongs to the TRPM (M for melastatin) subgroup of the family of transient receptor potential (TRP) channels. TRPM1 is a ∼180kDa protein localized to the somas and dendrites of both rod and cone metabotropic (ON) bipolar cells and colocalized with mGluR6 at the dendritic tips, where it plays an important role in light-regulated conductance in ON bipolar cells. Genetic lesions in the TRPM1 gene are associated with complete congenital stationary night blindness in human and TRPM1 knock-out mice have a no-b-wave (nob) electroretinogram phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM1
Applications Western Blot
Classification Monoclonal
Clone 545H5
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q2TV84
Gene Symbols Trpm1; CSNB1C; Ltrpc1; Mlsn1; MLSN1
Host Species Mouse
Immunogen Detergent stabilized, C-terminal 1D4 tag-tagged recombinant full-length mouse TRPM1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NM_001039104.2
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.