Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trpm7, Mouse, Clone: S74-25, Abnova™

Catalog No. 89112989 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-989 Supplier Abnova Corporation Supplier No. MAB6663
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Trpm7.

Specifications

Antigen Trpm7
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S74-25
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Trpm7.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1.0-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Trpm7
Gene Alias 2310022G15Rik/4833414K03Rik/5033407O22Rik/CHAK/CHAK1/Ltpr7/Ltrpc7/TRP-PLIK
Gene Symbols Trpm7
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to C-terminus amino acids 1817-1863 of mouse Trpm7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58800
Test Specificity Detects ∼220KDa. No cross-reactivity against TrpM6.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.