Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSH, Mouse, Clone: TSB1, Abnova™

Catalog No. 89111150 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-150 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-150 Supplier Abnova Corporation Supplier No. MAB2856

Mouse monoclonal antibody raised against native TSH.

Specifications

Antigen TSH
Applications ELISA
Classification Monoclonal
Clone TSB1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TSH.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM Tris, 150mM NaCl, pH 7.5 (0.05% sodium azide)
Gene Alias CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Native purified human pituitary gland TSH.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081/7252
Test Specificity This antibody specifically recognizes beta subunit of human TSH. No cross-react with human LH, FSH, HCG.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.