Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHB, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89110422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-422 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-422 Supplier Abnova Corporation Supplier No. MAB1413
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TSHB.

Thyrotropin-stimulating hormone (TSH) is a noncovalently linked glycoprotein heterodimer and is part of a family of pituitary hormones containing a common alpha subunit (TSHA; see MIM 118850) and a unique beta subunit that confers specificity.[supplied by OMIM

Specifications

Antigen TSHB
Applications ELISA
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TSHB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Gene TSHB
Gene Alias CHNG4/TSH-BETA
Gene Symbols TSHB
Host Species Mouse
Immunogen Native purified human TSHB.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Test Specificity This antibody reacts with human TSH and TSHB. Less than 0.1% cross-reactivity with human alpha-hCG, intact hCG, LH, FSH, growth hormone, or prolactin.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.