Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC11 Antibody (4211), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243771
Description
Ensure accurate, reproducible results in Immunocytochemistry/Immunofluorescence
TTC11 Monoclonal antibody specifically detects TTC11 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
TTC11 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
FIS1 | |
Full length human FIS1 Recombinant protein. | |
0.1 mL | |
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
51024 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
4211 | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction