Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA1B, Mouse, Clone: 17H11.F10, Abnova™

Catalog No. 89113364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-364 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-364 Supplier Abnova Corporation Supplier No. MAB8159
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TUBA1B.

Specifications

Antigen TUBA1B
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 17H11.F10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TUBA1B.
Dilution ELISA (1:300000) Western Blot (1:1000) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene TUBA1B
Gene Alias K-ALPHA-1
Gene Symbols TUBA1B
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus end of human TUBA1B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376
Test Specificity Cross reactivity will occur with all isoforms of alpha tubulin.
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.