Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB, Mouse, Clone: 37B, Abnova™

Catalog No. 89113887 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-887 Supplier Abnova Corporation Supplier No. MAB9929
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TUBB.

Specifications

Antigen TUBB
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 37B
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TUBB.
Dilution Western blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene TUBB
Gene Accession No. NM_178014.2
Gene Alias M40/MGC117247/MGC16435/OK/SW-cl.56/TUBB1/TUBB5
Gene Symbols TUBB
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 400-444 of human TUBB.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203068
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.