Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBG1, Mouse, Clone: TU-32, Abnova™

Catalog No. 89111516 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-516 Supplier Abnova Corporation Supplier No. MAB3661
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TUBG1.

This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7. [provided by RefSeq

Sequence: EYHAATRPDYISWGTQ

Specifications

Antigen TUBG1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TU-32
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TUBG1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TUBG1
Gene Alias TUBG/TUBGCP1
Gene Symbols TUBG1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 434-449 of human TUBG1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7283
Test Specificity This antibody recognizes an epitope (amino acids 434-449 in human) within C-terminus of TUBG1, a 48 KDa structural constituent of cytoskeleton and microtubule organizing center (MTOC).
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.