Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBG1, Mouse, Dyomics 647, Clone: TU-30, Abnova™

Catalog No. 89111845 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-845 Supplier Abnova Corporation Supplier No. MAB4476

Mouse monoclonal antibody raised against synthetic peptide of TUBG1.

This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7. [provided by RefSeq

Specifications

Antigen TUBG1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TU-30
Conjugate Dyomics 647
Description tubulin, gamma 1
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TUBG1
Gene Alias TUBG, TUBGCP1
Gene Symbols TUBG1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human TUBG1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7283
Test Specificity This antibody recognizes C-terminal peptide sequence EYHAATRPDYISWGTQ (aa 434-449) of gamma-tubulin, a 48kDa structural constituent of cytoskeleton and microtubule organizing center (MTOC).
Target Species Chicken, Human, Mouse, Plant, Pig, Protozoa, Rat
Content And Storage Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.