Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin-alpha/beta, Mouse, Clone: TU-10, Abnova™

Catalog No. 89135518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-518 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-518 Supplier Abnova Corporation Supplier No. MAB3825
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Tubulin-alpha/beta.

Specifications

Antigen Tubulin-alpha/beta
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TU-10
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Tubulin-alpha/beta.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Tubulin-alpha/beta from porcine brain microtubule proteins.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes alpha, beta-tubulin heterodimer, a basic intracellular structural unit of microtubules. Alpha- and beta-tubulins form approximately 100 KDa tubulin heterodimer, a globular protein that polymerizes to form microtubules.
Target Species Pig
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.