Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin-alpha, Mouse, Dyomics 547, Clone: TU-01, Abnova™

Catalog No. 89135893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-893 Supplier Abnova Corporation Supplier No. MAB5026

Mouse monoclonal antibody raised against native tubulin-alpha.

Specifications

Antigen Tubulin-alpha
Applications Immunocytochemistry
Classification Monoclonal
Clone TU-01
Conjugate Dyomics 547
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified porcine tubulin-alpha.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the defined epitope (aa 65-97) on N-terminal structural domain of alpha-tubulin.
Target Species Pig
Content And Storage Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.