Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin beta, Mouse, Clone: H3, Abnova™

Catalog No. 89136604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-604 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-604 Supplier Abnova Corporation Supplier No. MAB8770
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Tubulin beta.

Specifications

Antigen Tubulin beta
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone H3
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Tubulin beta.
Dilution Western Blot (1:1000) Immunofluorescence (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to human Tubulin beta.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chlamydomonas species, Drosophila, Frog, Human, Kangaroo, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.