Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-α-Tubulin, Clone: DM1A, Alexa Fluor 647,

Catalog No. 05829AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-829-AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-829-AF647 Supplier MilliporeSigma™ Supplier No. 05829AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects α-Tubulin clone: DM1A in Avian, Bovine, Gerbil, Guinea Pig, Human, Mouse, Porcine, Rat samples, and it is validated for Immunocytochemistry

Tubulin alpha-1 chain (UniProt P02552) is encoded by the TUBA1C (also known as TUBA1) gene (Gene ID 429035) in Gallus gallus (Chicken) species. Microtubules are cytoskeletal filaments composed of alpha-/beta-tubulin heterodimers that self-assemble head-to-tail to form protofilaments and laterally to form a hollow tube. Various post-translational modifications, including acetylation, polyglutamylation, polyglycylation, detyrosination, phosphorylation, and palmitoylation, can regulate the polymerization properties of tubulins and/or their interactions with microtubule associated proteins (MAPs) and motor proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Tubulin
Applications Immunocytochemistry
Classification Monoclonal
Clone DM1A
Conjugate Alexa Fluor 647
Formulation Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P02552
Gene Symbols TUBA1C; TUBA1
Host Species Mouse
Immunogen Purified chicken brain α-Tubulin. The epitope recognized by this antibody has been mapped to the C-terminal 25 amino acids (AALEKDYEEVGVDSVEGEGEEEGEE).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001292201.1
Target Species Avian, Bovine, Gerbil, Guinea Pig, Human, Mouse, Pig, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.